Gene Transcripts

Gene Symbol
Ceacam9
Entrez Gene ID
116711
Gene Name
carcinoembryonic antigen-related cell adhesion molecule 9
Gene Aliases
Chromosome Location
Chr. 1: 78659435 - 78663970 on Build Rnor_6.0
UniGene ID
Rn.81017
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053919.2 NP_446371.1 2 360
GenBank mRNA AF172446.1 - 2 362
FQ220766.1 - 2 360
FQ229599.1 - 2 354

View Full Product Details