Gene Transcripts

Gene Symbol
Cacng7
Entrez Gene ID
140728
Gene Name
calcium channel, voltage-dependent, gamma subunit 7
Gene Aliases
Chromosome Location
Chr. 1: 64374552 - 64405365 on Build Rnor_6.0
UniGene ID
Rn.81218
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080695.1 NP_542426.1 5 585
XM_006228013.2 XP_006228075.1 6 724
GenBank mRNA AF361345.1 - 5 585

View Full Product Details