Gene Transcripts

Gene Symbol
Tff1
Entrez Gene ID
117270
Gene Name
trefoil factor 1
Gene Aliases
Ps2
Chromosome Location
Chr. 20: 9892124 - 9895984 on Build Rnor_6.0
UniGene ID
Rn.10715
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057129.1 NP_476470.1 2 228
GenBank mRNA D83231.1 - 2 228

View Full Product Details