Gene Transcripts

Gene Symbol
Asb12
Entrez Gene ID
503446
Gene Name
ankyrin repeat and SOCS box-containing 12
Gene Aliases
Chromosome Location
Chr. X: 64714337 - 64800662 on Build Rnor_6.0
UniGene ID
Rn.8275
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037367.1 NP_001032444.1 4 1096
XM_006257044.2 XP_006257106.1 3 1049
GenBank mRNA BC107911.1 - 4 1096

View Full Product Details