Gene Transcripts
- Gene Symbol
- 1700108F19Rik
- Entrez Gene ID
- 73272
- Gene Name
- RIKEN cDNA 1700108F19 gene
- Gene Aliases
- 1700020I02Rik, BB065030
- Chromosome Location
- Chr. 14: 76677568 - 76687108 on Build GRCm38
- UniGene ID
- Mm.116804
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CF105082.1 |
- |
2 |
102 |
|
Chr. 14 - 76677605 - 76680330 [+] |
GenBankmRNA |
CN841323.1 |
- |
2 |
104 |
|
Chr. 14 - 76677607 - 76687107 [+] |
GenBankmRNA |
CB271207.1 |
- |
2 |
95 |
|
Chr. 14 - 76677612 - 76686890 [+] |
View Full Product Details