Gene Transcripts
- Gene Symbol
- 1700108F19Rik
- Entrez Gene ID
- 73272
- Gene Name
- RIKEN cDNA 1700108F19 gene
- Gene Aliases
- 1700020I02Rik, BB065030
- Chromosome Location
- Chr. 14: 76677568 - 76687108 on Build GRCm38
- UniGene ID
- Mm.116804
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015485.1 |
|
5 |
826 |
|
GenBank mRNA |
AK006165.1 |
- |
4 |
1386 |
|
AK007133.1 |
- |
5 |
815 |
|
View Full Product Details