Gene Transcripts
- Gene Symbol
- GOLGA8F
- Entrez Gene ID
- 100132565
- Gene Name
- golgin A8 family member F
- Gene Aliases
- Chromosome Location
- Chr. 15: 28378638 - 28391266 on Build GRCh38
- UniGene ID
- Hs.510812
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033351.1 |
|
18 |
3359 |
|
View Full Product Details