Gene Transcripts
- Gene Symbol
- CNTFR-AS1
- Entrez Gene ID
- 415056
- Gene Name
- CNTFR antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 9: 34568012 - 34583072 on Build GRCh38
- UniGene ID
- Hs.734229
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI460276.1 |
- |
2 |
481 |
|
Chr. 9 - 34568012 - 34582829 [+] |
GenBankmRNA |
BX092769.1 |
- |
2 |
468 |
|
Chr. 9 - 34568025 - 34582682 [+] |
View Full Product Details