Gene Transcripts

Gene Symbol
CNTFR-AS1
Entrez Gene ID
415056
Gene Name
CNTFR antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 9: 34568012 - 34583072 on Build GRCh38
UniGene ID
Hs.734229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024369.1 2 848
GenBank mRNA BC033987.1 - 3 960
BC038740.1 - 5 1281
BC042806.1 - 2 849

View Full Product Details