Gene Transcripts
- Gene Symbol
- Hipk2
- Entrez Gene ID
- 15258
- Gene Name
- homeodomain interacting protein kinase 2
- Gene Aliases
- 1110014O20Rik, B230339E18Rik, Stank
- Chromosome Location
- Chr. 6: 38687512 - 38876190 on Build GRCm38
- UniGene ID
- Mm.23790
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK003718.1 |
- |
1 |
1252 |
|
View Full Product Details