Gene Transcripts
- Gene Symbol
- 5430414B12Rik
- Entrez Gene ID
- 66788
- Gene Name
- RIKEN cDNA 5430414B12 gene
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK013463.1 |
- |
2 |
481 |
|
AK017306.1 |
- |
2 |
275 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY717118.1 |
- |
2 |
273 |
|
Chr. 1 - 191817600 - 191819429 [+] |
GenBankmRNA |
BE198858.1 |
- |
2 |
436 |
|
Chr. 1 - 191817437 - 191818918 [+] |
View Full Product Details