Gene Transcripts
- Gene Symbol
- Gm26604
- Entrez Gene ID
- 233045
- Gene Name
- predicted gene, 26604
- Gene Aliases
- C130030J05
- Chromosome Location
- Chr. 7: 29071642 - 29079906 on Build GRCm38
- UniGene ID
- Mm.226037
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK048019.1 |
- |
|
197 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB648634.1 |
- |
1, 2 |
194 |
|
Chr. 7 - 29071606 - 29080054 [+] |
View Full Product Details