Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003013.2 | NP_003004.1 | 3 | 896 | |
GenBank mRNA | AF311912.1 | - | 3 | 892 | |
AK075372.1 | - | 3 | 889 | ||
AK093922.1 | - | 4 | 864 | ||
AY359001.1 | - | 3 | 698 | ||
BC008666.1 | - | 3 | 876 | 3874262 |