Gene Transcripts
- Gene Symbol
- GGA1
- Entrez Gene ID
- 26088
- Gene Name
- golgi associated, gamma adaptin ear containing, ARF binding protein 1
- Gene Aliases
- Chromosome Location
- Chr. 22: 37607235 - 37633564 on Build GRCh38
- UniGene ID
- Hs.499158
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001001560.2 |
NP_001001560.1
|
1, 2 |
433 |
|
NM_001172687.1 |
NP_001166158.1
|
1, 2 |
433 |
|
NM_013365.4 |
NP_037497.1
|
1, 2 |
433 |
|
XM_005261517.3 |
XP_005261574.1
|
2, 3 |
607 |
|
XM_011530122.1 |
XP_011528424.1
|
2, 3 |
540 |
|
XM_011530124.1 |
XP_011528426.1
|
2, 3 |
607 |
|
XM_017028760.1 |
XP_016884249.1
|
2, 3 |
607 |
|
XM_017028762.1 |
XP_016884251.1
|
2, 3 |
607 |
|
XM_017028763.1 |
XP_016884252.1
|
1, 2 |
73 |
|
GenBank mRNA |
AF190862.1 |
- |
1, 2 |
61 |
|
AF218584.1 |
- |
1, 2 |
50 |
|
AF233521.1 |
- |
1, 2 |
68 |
|
AK075256.1 |
- |
1, 2 |
411 |
|
AK225157.1 |
- |
1, 2 |
67 |
|
AK290548.1 |
- |
1, 2 |
72 |
|
BC010917.1 |
- |
1, 2 |
77 |
|
BC029388.1 |
- |
1, 2 |
72 |
|
BC044629.1 |
- |
1, 2 |
62 |
5796877 |
CR456493.1 |
- |
1, 2 |
182 |
|
View Full Product Details