Gene Transcripts
- Gene Symbol
- Olfr1051
- Entrez Gene ID
- 404324
- Gene Name
- olfactory receptor 1051
- Gene Aliases
- GA_x5J8B7W3KVV-25848-25992, MOR187-5
- Chromosome Location
- Chr. 2: 86275559 - 86276485 on Build GRCm38
- UniGene ID
- Mm.377277
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207562.1 |
NP_997445.1
|
1 |
618 |
|
- Gene Symbol
- Olfr1049
- Entrez Gene ID
- 259018
- Gene Name
- olfactory receptor 1049
- Gene Aliases
- MOR187-1
- Chromosome Location
- Chr. 2: 86254765 - 86258320 on Build GRCm38
- UniGene ID
- Mm.223405
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_147016.2 |
NP_667227.2
|
1 |
618 |
|
View Full Product Details