Gene Transcripts

Gene Symbol
IQSEC3
Entrez Gene ID
440073
Gene Name
IQ motif and Sec7 domain 3
Gene Aliases
Chromosome Location
Chr. 12: 66794 - 178460 on Build GRCh38
UniGene ID
Hs.536319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC024764.1 - 1 1035

View Full Product Details