Gene Transcripts
- Gene Symbol
- FAM153A
- Entrez Gene ID
- 285596
- Gene Name
- family with sequence similarity 153 member A
- Gene Aliases
- NY-REN-7
- Chromosome Location
- Chr. 5: 177694247 - 177783518 on Build GRCh38
- UniGene ID
- Hs.368516
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173663.3 |
NP_775934.3
|
20 |
1677 |
|
GenBank mRNA |
AF155097.1 |
- |
20 |
1848 |
|
- Gene Symbol
- FAM153B
- Entrez Gene ID
- 202134
- Gene Name
- family with sequence similarity 153 member B
- Gene Aliases
- Chromosome Location
- Chr. 5: 176063709 - 176114798 on Build GRCh38
- UniGene ID
- Hs.368516
- Species
- Homo sapiens
View Full Product Details