Gene Transcripts
- Gene Symbol
- GOLGA8G
- Entrez Gene ID
- 283768
- Gene Name
- golgin A8 family member G
- Gene Aliases
- Chromosome Location
- Chr. 15: 28519611 - 28532997 on Build GRCh38
- UniGene ID
- Hs.169639
- Species
- Homo sapiens
- Gene Symbol
- GOLGA8CP
- Entrez Gene ID
- 729786
- Gene Name
- golgin A8 family member C, pseudogene
- Gene Aliases
- GOLGA8C
- Chromosome Location
- Chr. 15: 20562349 - 20575703 on Build GRCh38
- UniGene ID
- Hs.592025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027411.2 |
|
18 |
2628 |
|
- Gene Symbol
- GOLGA8DP
- Entrez Gene ID
- 100132979
- Gene Name
- golgin A8 family member D, pseudogene
- Gene Aliases
- GOLGA8D
- Chromosome Location
- Chr. 15: 23157340 - 23170783 on Build GRCh38
- UniGene ID
- Hs.454647
- Species
- Homo sapiens
- Gene Symbol
- GOLGA8EP
- Entrez Gene ID
- 390535
- Gene Name
- golgin A8 family member E, pseudogene
- Gene Aliases
- GOLGA8E
- Chromosome Location
- Chr. 15: 22424673 - 22438026 on Build GRCh38
- UniGene ID
- Hs.454647
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033350.1 |
|
18 |
2616 |
|
GenBank mRNA |
BC063309.1 |
- |
19 |
2590 |
|
DQ309037.1 |
- |
18 |
2590 |
|
- Gene Symbol
- GOLGA8F
- Entrez Gene ID
- 100132565
- Gene Name
- golgin A8 family member F
- Gene Aliases
- Chromosome Location
- Chr. 15: 28378638 - 28391266 on Build GRCh38
- UniGene ID
- Hs.510812
- Species
- Homo sapiens
- Gene Symbol
- ITSN2
- Entrez Gene ID
- 50618
- Gene Name
- intersectin 2
- Gene Aliases
- PRO2015, SH3D1B, SH3P18, SWA, SWAP
- Chromosome Location
- Chr. 2: 24202866 - 24360528 on Build GRCh38
- UniGene ID
- Hs.432562
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U61167.1 |
- |
8 |
1094 |
|
- Gene Symbol
- LOC645752
- Entrez Gene ID
- 645752
- Gene Name
- golgin A6 family member A pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 15: 77914217 - 77926846 on Build GRCh38
- UniGene ID
- Hs.546408
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027024.1 |
|
19 |
2525 |
|
GenBank mRNA |
AK294847.1 |
- |
3 |
820 |
|
AK308609.1 |
- |
4 |
861 |
|
View Full Product Details