Gene Transcripts
- Gene Symbol
- CTTNBP2
- Entrez Gene ID
- 83992
- Gene Name
- cortactin binding protein 2
- Gene Aliases
- C7orf8, CORTBP2, Orf4
- Chromosome Location
- Chr. 7: 117710648 - 117873507 on Build GRCh38
- UniGene ID
- Hs.592285
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033427.2 |
NP_219499.1
|
15, 16 |
3788 |
|
XM_005250634.4 |
XP_005250691.1
|
15, 16 |
3923 |
|
XM_005250635.3 |
XP_005250692.1
|
15, 16 |
3557 |
|
XM_011516615.2 |
XP_011514917.1
|
15, 16 |
6091 |
|
XM_011516616.2 |
XP_011514918.1
|
15, 16 |
3720 |
|
XM_017012706.1 |
XP_016868195.1
|
15, 16 |
3720 |
|
XM_017012708.1 |
XP_016868197.1
|
12, 13 |
3302 |
|
GenBank mRNA |
AB051545.1 |
- |
15, 16 |
3694 |
|
AF377960.1 |
- |
15, 16 |
3788 |
|
AK096565.1 |
- |
3, 4 |
325 |
|
BC106000.1 |
- |
15, 16 |
3755 |
30528261 |
View Full Product Details