Gene Transcripts

Gene Symbol
1700108F19Rik
Entrez Gene ID
73272
Gene Name
RIKEN cDNA 1700108F19 gene
Gene Aliases
1700020I02Rik, BB065030
Chromosome Location
Chr. 14: 76677568 - 76687108 on Build GRCm38
UniGene ID
Mm.116804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK006165.1 - 3 975
AK007133.1 - 4 405
AK076702.1 - 4 374
BY707214.1 - 4 403

View Full Product Details