Gene Transcripts
- Gene Symbol
- Spns1
- Entrez Gene ID
- 73658
- Gene Name
- spinster homolog 1
- Gene Aliases
- 2210013K02Rik, Spin1, Spinl
- Chromosome Location
- Chr. 7: 126370060 - 126377934 on Build GRCm38
- UniGene ID
- Mm.11112
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF212372.1 |
- |
4 |
913 |
|
View Full Product Details