Gene Transcripts
- Gene Symbol
- TRPV4
- Entrez Gene ID
- 59341
- Gene Name
- transient receptor potential cation channel subfamily V member 4
- Gene Aliases
- BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC
- Chromosome Location
- Chr. 12: 109783087 - 109833407 on Build GRCh38
- UniGene ID
- Hs.506713
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001177428.1 |
NP_001170899.1
|
2, 3 |
588 |
|
NM_001177431.1 |
NP_001170902.1
|
3, 4 |
455 |
|
NM_001177433.1 |
NP_001170904.1
|
2, 3 |
588 |
|
NM_021625.4 |
NP_067638.3
|
3, 4 |
646 |
|
NM_147204.2 |
NP_671737.1
|
2, 3 |
588 |
|
XM_005253918.1 |
XP_005253975.1
|
3, 4 |
1006 |
|
XM_011538630.2 |
XP_011536932.2
|
3, 4 |
1749 |
|
XM_011538631.2 |
XP_011536933.2
|
3, 4 |
1750 |
|
XM_011538632.2 |
XP_011536934.2
|
3, 4 |
1750 |
|
XM_011538633.2 |
XP_011536935.2
|
3, 4 |
1750 |
|
XM_011538634.2 |
XP_011536936.2
|
3, 4 |
1751 |
|
XM_011538635.2 |
XP_011536937.1
|
3, 4 |
1753 |
|
XM_011538636.2 |
XP_011536938.1
|
3, 4 |
1754 |
|
XM_017019774.1 |
XP_016875263.1
|
3, 4 |
1379 |
|
GenBank mRNA |
AB032427.1 |
- |
2, 3 |
652 |
|
AB073669.1 |
- |
2, 3 |
557 |
|
AF258465.1 |
- |
2, 3 |
557 |
|
AF279673.1 |
- |
2, 3 |
353 |
|
AJ296305.1 |
- |
3, 4 |
646 |
|
AL834192.1 |
- |
3, 4 |
618 |
|
BC117426.1 |
- |
3, 4 |
640 |
40125977 |
BC143307.1 |
- |
3, 4 |
640 |
|
BC143315.1 |
- |
3, 4 |
640 |
9051820 |
DQ059644.1 |
- |
2, 3 |
557 |
|
DQ059645.1 |
- |
3, 4 |
455 |
|
DQ059646.1 |
- |
2, 3 |
557 |
|
View Full Product Details