Gene Transcripts

Gene Symbol
ARFIP2
Entrez Gene ID
23647
Gene Name
ADP ribosylation factor interacting protein 2
Gene Aliases
POR1
Chromosome Location
Chr. 11: 6474683 - 6481479 on Build GRCh38
UniGene ID
Hs.732203
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242854.1 NP_001229783.1 8 2420
NM_001242855.1 NP_001229784.1 7 2180
NM_001242856.1 NP_001229785.1 6 2061
NM_012402.3 NP_036534.1 8 2281
XM_005252840.4 XP_005252897.1 8 2185
GenBank mRNA AK026390.1 - 8 2113

View Full Product Details