Gene Transcripts

Gene Symbol
Dsp
Entrez Gene ID
109620
Gene Name
desmoplakin
Gene Aliases
2300002E22Rik, 5730453H04Rik, AA407887, AA407888, AW109828, DP, rul
Chromosome Location
Chr. 13: 38151084 - 38198577 on Build GRCm38
UniGene ID
Mm.355327
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023842.2 NP_076331.2 24 8220
GenBank mRNA AK077574.1 - 1 722

View Full Product Details