Gene Transcripts

Gene Symbol
Myo5a
Entrez Gene ID
25017
Gene Name
myosin VA
Gene Aliases
D, Dop, Myh12
Chromosome Location
Chr. 8: 82038966 - 82156507 on Build Rnor_6.0
UniGene ID
Rn.44865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022178.1 NP_071514.1 2760
GenBank mRNA AB035736.1 - 2760

View Full Product Details