Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
Chromosome Location
Chr. 3: 14489111 - 14602996 on Build GRCh38
UniGene ID
Hs.517819
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080423.3 NP_001073892.3 24 6103
GenBank mRNA AB051506.2 - 24 6089

View Full Product Details