Gene Transcripts

Gene Symbol
DSC1
Entrez Gene ID
1823
Gene Name
desmocollin 1
Gene Aliases
CDHF1, DG2/DG3
Chromosome Location
Chr. 18: 31129236 - 31162856 on Build GRCh38
UniGene ID
Hs.567260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004948.3 NP_004939.1 9 1370
NM_024421.2 NP_077739.1 9 1370
GenBank mRNA X72925.1 - 9 1370
Z34522.1 - 9 1298

View Full Product Details