Gene Transcripts
- Gene Symbol
- MYO7B
- Entrez Gene ID
- 4648
- Gene Name
- myosin VIIB
- Gene Aliases
- Chromosome Location
- Chr. 2: 127535689 - 127637728 on Build GRCh38
- UniGene ID
- Hs.154578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074183.1 |
- |
15 |
2587 |
|
View Full Product Details