Gene Transcripts
- Gene Symbol
- APBB2
- Entrez Gene ID
- 323
- Gene Name
- amyloid beta precursor protein binding family B member 2
- Gene Aliases
- FE65L, FE65L1
- Chromosome Location
- Chr. 4: 40810027 - 41216714 on Build GRCh38
- UniGene ID
- Hs.479602
- Species
- Homo sapiens
- Gene Symbol
- ABCA5
- Entrez Gene ID
- 23461
- Gene Name
- ATP binding cassette subfamily A member 5
- Gene Aliases
- ABC13, EST90625
- Chromosome Location
- Chr. 17: 69244435 - 69327182 on Build GRCh38
- UniGene ID
- Hs.731824
- Species
- Homo sapiens
- Gene Symbol
- AXL
- Entrez Gene ID
- 558
- Gene Name
- AXL receptor tyrosine kinase
- Gene Aliases
- ARK, JTK11, Tyro7, UFO
- Chromosome Location
- Chr. 19: 41219199 - 41261767 on Build GRCh38
- UniGene ID
- Hs.590970
- Species
- Homo sapiens
- Gene Symbol
- C17orf105
- Entrez Gene ID
- 284067
- Gene Name
- chromosome 17 open reading frame 105
- Gene Aliases
- Chromosome Location
- Chr. 17: 43778391 - 43784842 on Build GRCh38
- UniGene ID
- Hs.252707
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125817.1 |
- |
6 |
3262 |
|
- Gene Symbol
- CDKL1
- Entrez Gene ID
- 8814
- Gene Name
- cyclin dependent kinase like 1
- Gene Aliases
- KKIALRE, P42
- Chromosome Location
- Chr. 14: 50329404 - 50397404 on Build GRCh38
- UniGene ID
- Hs.280881
- Species
- Homo sapiens
- Gene Symbol
- COL6A4P2
- Entrez Gene ID
- 646300
- Gene Name
- collagen type VI alpha 4 pseudogene 2
- Gene Aliases
- COL6A4
- Chromosome Location
- Chr. 3: 130212820 - 130273806 on Build GRCh38
- UniGene ID
- Hs.170475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027898.1 |
|
23 |
3206 |
|
- Gene Symbol
- FBXW2
- Entrez Gene ID
- 26190
- Gene Name
- F-box and WD repeat domain containing 2
- Gene Aliases
- FBW2, Fwd2, Md6
- Chromosome Location
- Chr. 9: 120756976 - 120793471 on Build GRCh38
- UniGene ID
- Hs.494985
- Species
- Homo sapiens
- Gene Symbol
- FER1L4
- Entrez Gene ID
- 80307
- Gene Name
- fer-1 like family member 4, pseudogene
- Gene Aliases
- C20orf124
- Chromosome Location
- Chr. 20: 35558737 - 35607562 on Build GRCh38
- UniGene ID
- Hs.72222
- Species
- Homo sapiens
- Gene Symbol
- GGCX
- Entrez Gene ID
- 2677
- Gene Name
- gamma-glutamyl carboxylase
- Gene Aliases
- VKCFD1
- Chromosome Location
- Chr. 2: 85544720 - 85561534 on Build GRCh38
- UniGene ID
- Hs.77719
- Species
- Homo sapiens
- Gene Symbol
- ING5
- Entrez Gene ID
- 84289
- Gene Name
- inhibitor of growth family member 5
- Gene Aliases
- p28ING5
- Chromosome Location
- Chr. 2: 241687017 - 241729481 on Build GRCh38
- UniGene ID
- Hs.529172
- Species
- Homo sapiens
- Gene Symbol
- LIN54
- Entrez Gene ID
- 132660
- Gene Name
- lin-54 DREAM MuvB core complex component
- Gene Aliases
- CXCDC1, JC8.6, MIP120, TCX1
- Chromosome Location
- Chr. 4: 82924603 - 83012951 on Build GRCh38
- UniGene ID
- Hs.96952
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX108765.1 |
- |
1 |
426 |
|
- Gene Symbol
- LIX1L
- Entrez Gene ID
- 128077
- Gene Name
- limb and CNS expressed 1 like
- Gene Aliases
- Chromosome Location
- Chr. 1: 145933423 - 145958038 on Build GRCh38
- UniGene ID
- Hs.740619
- Species
- Homo sapiens
- Gene Symbol
- LLGL2
- Entrez Gene ID
- 3993
- Gene Name
- LLGL2, scribble cell polarity complex component
- Gene Aliases
- HGL, Hugl-2, LGL2
- Chromosome Location
- Chr. 17: 75525080 - 75575209 on Build GRCh38
- UniGene ID
- Hs.514477
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125563.1 |
- |
1 |
472 |
|
- Gene Symbol
- LYRM4
- Entrez Gene ID
- 57128
- Gene Name
- LYR motif containing 4
- Gene Aliases
- C6orf149, CGI-203, COXPD19, ISD11
- Chromosome Location
- Chr. 6: 5031029 - 5260950 on Build GRCh38
- UniGene ID
- Hs.387755
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_104417.1 |
|
3 |
573 |
|
GenBank mRNA |
AK309458.1 |
- |
2 |
272 |
|
- Gene Symbol
- MS4A10
- Entrez Gene ID
- 341116
- Gene Name
- membrane spanning 4-domains A10
- Gene Aliases
- CD20L7, MS4A9
- Chromosome Location
- Chr. 11: 60785332 - 60801305 on Build GRCh38
- UniGene ID
- Hs.591956
- Species
- Homo sapiens
- Gene Symbol
- NFKBID
- Entrez Gene ID
- 84807
- Gene Name
- NFKB inhibitor delta
- Gene Aliases
- IkappaBNS, TA-NFKBH
- Chromosome Location
- Chr. 19: 35887630 - 35902301 on Build GRCh38
- UniGene ID
- Hs.466531
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK126825.1 |
- |
4 |
725 |
|
- Gene Symbol
- OGFOD1
- Entrez Gene ID
- 55239
- Gene Name
- 2-oxoglutarate and iron dependent oxygenase domain containing 1
- Gene Aliases
- TPA1
- Chromosome Location
- Chr. 16: 56451480 - 56477495 on Build GRCh38
- UniGene ID
- Hs.231883
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB046832.1 |
- |
13 |
3792 |
|
- Gene Symbol
- RWDD2A
- Entrez Gene ID
- 112611
- Gene Name
- RWD domain containing 2A
- Gene Aliases
- RWDD2, dJ747H23.2
- Chromosome Location
- Chr. 6: 83193313 - 83198935 on Build GRCh38
- UniGene ID
- Hs.423336
- Species
- Homo sapiens
- Gene Symbol
- SIGLEC11
- Entrez Gene ID
- 114132
- Gene Name
- sialic acid binding Ig like lectin 11
- Gene Aliases
- Chromosome Location
- Chr. 19: 49948993 - 49961172 on Build GRCh38
- UniGene ID
- Hs.661852
- Species
- Homo sapiens
- Gene Symbol
- SMIM11A
- Entrez Gene ID
- 54065
- Gene Name
- small integral membrane protein 11A
- Gene Aliases
- C21orf51, FAM165B, SMIM11
- Chromosome Location
- Chr. 21: 34375450 - 34402777 on Build GRCh38
- UniGene ID
- Hs.656195
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BM989806.1 |
- |
1 |
270 |
|
- Gene Symbol
- TPGS2
- Entrez Gene ID
- 25941
- Gene Name
- tubulin polyglutamylase complex subunit 2
- Gene Aliases
- C18orf10, HMFN0601, L17, PGs2
- Chromosome Location
- Chr. 18: 36777647 - 36829393 on Build GRCh38
- UniGene ID
- Hs.436636
- Species
- Homo sapiens
- Gene Symbol
- TRPM7
- Entrez Gene ID
- 54822
- Gene Name
- transient receptor potential cation channel subfamily M member 7
- Gene Aliases
- ALSPDC, CHAK, CHAK1, LTRPC7, LTrpC-7, TRP-PLIK
- Chromosome Location
- Chr. 15: 50557158 - 50686835 on Build GRCh38
- UniGene ID
- Hs.512894
- Species
- Homo sapiens
- Gene Symbol
- URGCP
- Entrez Gene ID
- 55665
- Gene Name
- upregulator of cell proliferation
- Gene Aliases
- URG4
- Chromosome Location
- Chr. 7: 43875894 - 43926411 on Build GRCh38
- UniGene ID
- Hs.663312
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832134.1 |
- |
1 |
1296 |
|
- Gene Symbol
- USF3
- Entrez Gene ID
- 205717
- Gene Name
- upstream transcription factor family member 3
- Gene Aliases
- KIAA2018
- Chromosome Location
- Chr. 3: 113648385 - 113696657 on Build GRCh38
- UniGene ID
- Hs.632570
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX537728.1 |
- |
1 |
3982 |
|
View Full Product Details