Gene Transcripts

Gene Symbol
MAGIX
Entrez Gene ID
79917
Gene Name
MAGI family member, X-linked
Gene Aliases
JM10, PDZX
Chromosome Location
Chr. X: 49162569 - 49168157 on Build GRCh38
UniGene ID
Hs.193170
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005278064.4 XP_005278121.1 2 233
XM_005278065.3 XP_005278122.1 2 321
XM_011543985.2 XP_011542287.1 3 454
XM_011543986.2 XP_011542288.1 2 208
XM_011543987.2 XP_011542289.1 2 229
GenBank mRNA AK025340.1 - 2 155
AK096058.1 - 2 1395
DQ884401.1 - 1 76

View Full Product Details