Gene Transcripts

Gene Symbol
Magix
Entrez Gene ID
54634
Gene Name
MAGI family member, X-linked
Gene Aliases
Pdzx, Sfc18
Chromosome Location
Chr. X: 7673166 - 7681251 on Build GRCm38
UniGene ID
Mm.67912
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018832.2 NP_061302.2 4 386
GenBank mRNA AF229645.1 - 4 406
AK077898.1 - 4 353
AK158091.1 - 4 326
BC099459.1 - 4 386 30847792
BC132580.1 - 4 325 40130857
BC132582.1 - 4 325 40130859

View Full Product Details