Gene Transcripts

Gene Symbol
Marveld3
Entrez Gene ID
73608
Gene Name
MARVEL (membrane-associating) domain containing 3
Gene Aliases
1810006A16Rik, AI642133, Marvd3, Mrvldc3
Chromosome Location
Chr. 8: 109947909 - 109962205 on Build GRCm38
UniGene ID
Mm.86833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028584.3 NP_082860.2 3 1658
GenBank mRNA AK007346.1 - 3 1579
BC038647.1 - 1 118

View Full Product Details