Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177428.1 | NP_001170899.1 | 6 | 1303 | |
NM_001177431.1 | NP_001170902.1 | 8 | 1311 | ||
NM_001177433.1 | NP_001170904.1 | 5 | 1123 | ||
NM_021625.4 | NP_067638.3 | 8 | 1502 | ||
NM_147204.2 | NP_671737.1 | 6 | 1264 | ||
XM_005253918.1 | XP_005253975.1 | 8 | 1862 | ||
XM_011538630.2 | XP_011536932.2 | 8 | 2605 | ||
XM_011538631.2 | XP_011536933.2 | 7 | 2465 | ||
XM_011538632.2 | XP_011536934.2 | 7 | 2426 | ||
XM_011538633.2 | XP_011536935.2 | 6 | 2285 | ||
XM_011538634.2 | XP_011536936.2 | 8 | 2607 | ||
XM_011538635.2 | XP_011536937.1 | 8 | 2609 | ||
XM_011538636.2 | XP_011536938.1 | 8 | 2610 | ||
XM_017019774.1 | XP_016875263.1 | 8 | 2235 | ||
GenBank mRNA | AB032427.1 | - | 7 | 1508 | |
AB073669.1 | - | 6 | 1233 | ||
AB100308.1 | - | 7 | 1413 | ||
AF258465.1 | - | 7 | 1413 | ||
AF263523.1 | - | 7 | 1413 | ||
AF279673.1 | - | 7 | 1209 | ||
AJ296305.1 | - | 8 | 1502 | ||
AL834192.1 | - | 8 | 1473 | ||
BC117426.1 | - | 8 | 1496 | 40125977 | |
BC143307.1 | - | 8 | 1496 | ||
BC143315.1 | - | 8 | 1496 | 9051820 | |
DQ059644.1 | - | 6 | 1272 | ||
DQ059645.1 | - | 8 | 1311 | ||
DQ059646.1 | - | 5 | 1092 |