Gene Transcripts

Gene Symbol
Gabpb1
Entrez Gene ID
14391
Gene Name
GA repeat binding protein, beta 1
Gene Aliases
BABPB2, E4TF1, E4TF1-47, E4TF1-53, E4Tf1B, GABPB, GABPB-1, GABPB-2, GABPB1-1, GABPB1-2, NRF2B1, NRF2B2
Chromosome Location
Chr. 2: 126628907 - 126676677 on Build GRCm38
UniGene ID
Mm.293266
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271467.1 NP_001258396.1 9 1314
NM_001271468.1 NP_001258397.1 8 1219
NM_001271470.1 NP_001258399.1 9 1381
NM_010249.2 NP_034379.1 8 1219
NM_207669.2 NP_997552.1 8 1216
GenBank mRNA AI324106.1 - 1 234
AK052828.1 - 1195
AK139710.1 - 9 1381
AK140222.1 - 9 1267
AK140907.1 - 8 1127
AK146373.1 - 8 1083
AW492609.1 - 1 239
AY282800.1 - 5 676
AY282803.1 - 5 676
AY282804.1 - 6 758
AY282805.1 - 5 759
BC013558.1 - 8 1110 3592119
M74516.1 - 8 1111
M74517.1 - 8 1113

View Full Product Details