Gene Transcripts
- Gene Symbol
- LACTB2-AS1
- Entrez Gene ID
- 286190
- Gene Name
- LACTB2 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 8: 70608577 - 70663279 on Build GRCh38
- UniGene ID
- Hs.662204
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038881.1 |
|
4 |
2636 |
|
GenBank mRNA |
AL833348.1 |
- |
4 |
2636 |
|
View Full Product Details