Gene Transcripts
- Gene Symbol
- Gm29856
- Entrez Gene ID
- 102631545
- Gene Name
- predicted gene, 29856
- Gene Aliases
- Chromosome Location
- Chr. 1: 177457496 - 177461562 on Build GRCm38
- UniGene ID
- Mm.127605
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082514.1 |
- |
3 |
560 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB651099.1 |
- |
3 |
557 |
|
Chr. 1 - 177458969 - 177460617 [+] |
View Full Product Details