Gene Transcripts
- Gene Symbol
- Smg8
- Entrez Gene ID
- 287596
- Gene Name
- SMG8 nonsense mediated mRNA decay factor
- Gene Aliases
- RGD1306862
- Chromosome Location
- Chr. 10: 74379224 - 74388846 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_213430.7 |
XP_213430.2
|
1 |
2274 |
|
View Full Product Details