Gene Transcripts
- Gene Symbol
- GOLGA8F
- Entrez Gene ID
- 100132565
- Gene Name
- golgin A8 family member F
- Gene Aliases
- Chromosome Location
- Chr. 15: 28378638 - 28391266 on Build GRCh38
- UniGene ID
- Hs.510812
- Species
- Homo sapiens
- Gene Symbol
- GOLGA8CP
- Entrez Gene ID
- 729786
- Gene Name
- golgin A8 family member C, pseudogene
- Gene Aliases
- GOLGA8C
- Chromosome Location
- Chr. 15: 20562349 - 20575703 on Build GRCh38
- UniGene ID
- Hs.592025
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027411.2 |
|
10 |
1221 |
|
- Gene Symbol
- GOLGA8DP
- Entrez Gene ID
- 100132979
- Gene Name
- golgin A8 family member D, pseudogene
- Gene Aliases
- GOLGA8D
- Chromosome Location
- Chr. 15: 23157340 - 23170783 on Build GRCh38
- UniGene ID
- Hs.454647
- Species
- Homo sapiens
- Gene Symbol
- GOLGA8EP
- Entrez Gene ID
- 390535
- Gene Name
- golgin A8 family member E, pseudogene
- Gene Aliases
- GOLGA8E
- Chromosome Location
- Chr. 15: 22424673 - 22438026 on Build GRCh38
- UniGene ID
- Hs.454647
- Species
- Homo sapiens
- Gene Symbol
- GOLGA8G
- Entrez Gene ID
- 283768
- Gene Name
- golgin A8 family member G
- Gene Aliases
- Chromosome Location
- Chr. 15: 28519611 - 28532997 on Build GRCh38
- UniGene ID
- Hs.169639
- Species
- Homo sapiens
- Gene Symbol
- GOLGA8S
- Entrez Gene ID
- 653061
- Gene Name
- golgin A8 family member S
- Gene Aliases
- Chromosome Location
- Chr. 15: 23354748 - 23368324 on Build GRCh38
- UniGene ID
- Hs.528454
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038843.1 |
|
10 |
1195 |
|
View Full Product Details