Gene Transcripts
- Gene Symbol
- APOBEC3F
- Entrez Gene ID
- 200316
- Gene Name
- apolipoprotein B mRNA editing enzyme catalytic subunit 3F
- Gene Aliases
- A3F, ARP8, BK150C2.4.MRNA, KA6
- Chromosome Location
- Chr. 22: 39020454 - 39055972 on Build GRCh38
- UniGene ID
- Hs.659809
- Species
- Homo sapiens
- Gene Symbol
- ZNF253
- Entrez Gene ID
- 56242
- Gene Name
- zinc finger protein 253
- Gene Aliases
- BMZF-1, BMZF1, ZNF411
- Chromosome Location
- Chr. 19: 19865905 - 19893484 on Build GRCh38
- UniGene ID
- Hs.301059
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK074757.1 |
- |
6 |
4306 |
|
View Full Product Details