Gene Transcripts
- Gene Symbol
- SUMF2
- Entrez Gene ID
- 25870
- Gene Name
- sulfatase modifying factor 2
- Gene Aliases
- pFGE
- Chromosome Location
- Chr. 7: 56064007 - 56080672 on Build GRCh38
- UniGene ID
- Hs.279696
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001042469.1 |
NP_001035934.2
|
6, 7 |
717 |
|
NM_001042470.1 |
NP_001035935.2
|
4, 5 |
510 |
|
NM_001146333.1 |
NP_001139805.1
|
6, 7 |
605 |
|
NM_015411.2 |
NP_056226.2
|
7, 8 |
762 |
|
XM_011515254.2 |
XP_011513556.1
|
7, 8 |
762 |
|
XM_017011939.1 |
XP_016867428.1
|
7, 8 |
762 |
|
XM_017011940.1 |
XP_016867429.1
|
6, 7 |
934 |
|
GenBank mRNA |
AK075477.1 |
- |
7, 8 |
721 |
|
AK297042.1 |
- |
6, 7 |
633 |
|
AK297793.1 |
- |
7, 8 |
579 |
|
AK298605.1 |
- |
5, 6 |
495 |
|
AK300488.1 |
- |
6, 7 |
564 |
|
AK301627.1 |
- |
4, 5 |
427 |
|
AL050037.1 |
- |
5, 6 |
483 |
|
AY323911.1 |
- |
7, 8 |
674 |
|
AY359103.1 |
- |
7, 8 |
692 |
|
BC000224.1 |
- |
6, 7 |
497 |
|
BC006159.1 |
- |
7, 8 |
660 |
|
BC065222.1 |
- |
4, 5 |
398 |
|
BC084539.1 |
- |
7, 8 |
692 |
6599080 |
BC111092.1 |
- |
7, 8 |
715 |
40068651 |
BM423367.1 |
- |
7, 8 |
664 |
|
BP256067.1 |
- |
5, 6 |
404 |
|
CF131206.1 |
- |
7, 8 |
723 |
|
CR936749.1 |
- |
6, 7 |
652 |
|
CR936757.1 |
- |
7, 8 |
705 |
|
DA917807.1 |
- |
4, 5 |
479 |
|
DB010869.1 |
- |
4, 5 |
428 |
|
KU178583.1 |
- |
6, 7 |
587 |
|
KU178584.1 |
- |
5, 6 |
542 |
|
KU178585.1 |
- |
3, 4 |
335 |
|
View Full Product Details