Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026156.2 | NP_080432.1 | 11 | 1430 | |
GenBank mRNA | AK002890.1 | - | 11 | 1431 | |
AK031533.1 | - | 1517 | |||
AK087952.1 | - | 8 | 1693 | ||
AK132434.1 | - | 11 | 1444 | ||
AK145925.1 | - | 11 | 1432 | ||
BC021341.1 | - | 11 | 1428 | 5066146 |