Gene Transcripts

Gene Symbol
Setd1b
Entrez Gene ID
208043
Gene Name
SET domain containing 1B
Gene Aliases
AA516740, BC035291, KMT2G, mKIAA1076
Chromosome Location
Chr. 5: 123140787 - 123168634 on Build GRCm38
UniGene ID
Mm.250391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040398.2 NP_001035488.2 8, 9 3224
GenBank mRNA BC038367.1 - 2, 3 302

View Full Product Details