Gene Transcripts
- Gene Symbol
- CAMK1G
- Entrez Gene ID
- 57172
- Gene Name
- calcium/calmodulin dependent protein kinase IG
- Gene Aliases
- CLICK3, CLICKIII, VWS1, dJ272L16.1
- Chromosome Location
- Chr. 1: 209583700 - 209613939 on Build GRCh38
- UniGene ID
- Hs.199068
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020439.2 |
NP_065172.1
|
2, 3 |
204 |
|
XM_017001866.1 |
XP_016857355.1
|
2, 3 |
347 |
|
XM_017001867.1 |
XP_016857356.1
|
2, 3 |
187 |
|
GenBank mRNA |
AF428261.1 |
- |
2, 3 |
174 |
|
AK095713.1 |
- |
2, 3 |
347 |
|
AK289675.1 |
- |
2, 3 |
187 |
|
AL049688.1 |
- |
1, 2 |
119 |
|
AY212935.1 |
- |
1, 2 |
110 |
|
BC032787.1 |
- |
2, 3 |
171 |
5179957 |
BG715920.1 |
- |
2, 3 |
208 |
|
View Full Product Details