Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282669.1 | NP_001269598.1 | 3 | 686 | |
NM_001320132.1 | NP_001307061.1 | 3 | 783 | ||
NM_001320136.1 | NP_001307065.1 | 3 | 664 | ||
NM_004402.3 | NP_004393.1 | 3 | 614 | ||
NR_104222.1 | 3 | 636 | |||
NR_135150.1 | 3 | 614 | |||
NR_135151.1 | 4 | 701 | |||
NR_135152.1 | 3 | 636 | |||
XM_011540865.2 | XP_011539167.1 | 3 | 666 | ||
XM_017000499.1 | XP_016855988.1 | 3 | 666 | ||
XM_017000500.1 | XP_016855989.1 | 3 | 421 | ||
GenBank mRNA | AB013918.1 | - | 3 | 277 | |
AB028911.1 | - | 3 | 408 | ||
AB028912.1 | - | 3 | 577 | ||
AB028913.1 | - | 4 | 495 | ||
AF039210.1 | - | 3 | 277 | ||
AF064019.1 | - | 3 | 408 | ||
AF409060.1 | - | 5 | 288 | ||
AF409061.1 | - | 3 | 303 | ||
AF409062.1 | - | 3 | 331 | ||
AK290877.1 | - | 3 | 405 | ||
AK303065.1 | - | 3 | 493 | ||
AK309316.1 | - | 3 | 396 | ||
BC032827.1 | - | 3 | 448 | ||
BC048797.1 | - | 3 | 600 | 5204996 |