Gene Transcripts
- Gene Symbol
- IL6R
- Entrez Gene ID
- 3570
- Gene Name
- interleukin 6 receptor
- Gene Aliases
- CD126, IL-6R-1, IL-6RA, IL6Q, IL6RA, IL6RQ, gp80
- Chromosome Location
- Chr. 1: 154405193 - 154469450 on Build GRCh38
- UniGene ID
- Hs.135087
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000565.3 |
NP_000556.1
|
3, 4 |
897 |
|
NM_001206866.1 |
NP_001193795.1
|
3, 4 |
897 |
|
NM_181359.2 |
NP_852004.1
|
3, 4 |
897 |
|
XM_005245139.1 |
XP_005245196.1
|
3, 4 |
856 |
|
XM_005245140.2 |
XP_005245197.1
|
3, 4 |
856 |
|
XM_006711298.1 |
XP_006711361.1
|
3, 4 |
856 |
|
XM_006711299.3 |
XP_006711362.1
|
3, 4 |
856 |
|
XM_017001199.1 |
XP_016856688.1
|
3, 4 |
856 |
|
XM_017001200.1 |
XP_016856689.1
|
3, 4 |
856 |
|
XM_017001201.1 |
XP_016856690.1
|
3, 4 |
856 |
|
GenBank mRNA |
AK223582.1 |
- |
3, 4 |
718 |
|
AK293013.1 |
- |
3, 4 |
511 |
|
AK312730.1 |
- |
3, 4 |
508 |
|
BC035327.1 |
- |
3, 4 |
472 |
|
BC089410.1 |
- |
3, 4 |
867 |
3078506 |
BC132684.1 |
- |
3, 4 |
710 |
40146706 |
BC132686.1 |
- |
3, 4 |
710 |
40146708 |
X12830.1 |
- |
3, 4 |
897 |
|
X58298.1 |
- |
3, 4 |
511 |
|
View Full Product Details