Gene Transcripts
- Gene Symbol
- ME2
- Entrez Gene ID
- 4200
- Gene Name
- malic enzyme 2
- Gene Aliases
- ODS1
- Chromosome Location
- Chr. 18: 50878604 - 50949795 on Build GRCh38
- UniGene ID
- Hs.233119
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M55905.1 |
- |
11, 12 |
1252 |
|
View Full Product Details