Gene Transcripts
- Gene Symbol
- FXYD5
- Entrez Gene ID
- 53827
- Gene Name
- FXYD domain containing ion transport regulator 5
- Gene Aliases
- DYSAD, HSPC113, IWU1, KCT1, OIT2, PRO6241, RIC
- Chromosome Location
- Chr. 19: 35154722 - 35169885 on Build GRCh38
- UniGene ID
- Hs.333418
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001164605.1 |
NP_001158077.1
|
6, 7 |
474 |
|
NM_001320912.1 |
NP_001307841.1
|
6, 7 |
471 |
|
NM_001320913.1 |
NP_001307842.1
|
5, 6 |
378 |
|
NM_014164.5 |
NP_054883.3
|
6, 7 |
471 |
|
NM_144779.2 |
NP_659003.1
|
6, 7 |
484 |
|
NR_028406.1 |
|
5, 6 |
1165 |
|
GenBank mRNA |
AA044211.1 |
- |
6, 7 |
413 |
|
AB072911.1 |
- |
6, 7 |
450 |
|
AF161462.1 |
- |
5, 6 |
394 |
|
AF177940.1 |
- |
1, 2 |
212 |
|
AK304012.1 |
- |
6, 7 |
463 |
|
AY157582.1 |
- |
5, 6 |
383 |
|
AY358991.1 |
- |
6, 7 |
421 |
|
BC009642.2 |
- |
5, 6 |
830 |
3900880 |
BC013834.1 |
- |
6, 7 |
425 |
|
BC018798.1 |
- |
5, 6 |
1161 |
|
BG025158.1 |
- |
6, 7 |
525 |
|
BM550889.1 |
- |
6, 7 |
406 |
|
BM839241.1 |
- |
|
471 |
|
BQ440098.1 |
- |
6, 7 |
484 |
|
BQ941279.1 |
- |
6, 7 |
416 |
|
BT007343.1 |
- |
5, 6 |
383 |
|
BU164524.1 |
- |
6, 7 |
474 |
|
BU178888.1 |
- |
6, 7 |
469 |
|
View Full Product Details