Gene Transcripts

Gene Symbol
PDSS2
Entrez Gene ID
57107
Gene Name
prenyl (decaprenyl) diphosphate synthase, subunit 2
Gene Aliases
C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1
Chromosome Location
Chr. 6: 107152557 - 107459683 on Build GRCh38
UniGene ID
Hs.745008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020381.3 NP_065114.3 1 526
XM_011535956.2 XP_011534258.1 1 624
XM_011535957.2 XP_011534259.1 1 634
XM_011535958.2 XP_011534260.1 1 625
XM_011535959.2 XP_011534261.1 1 599
XM_011535961.2 XP_011534263.1 1 546
XM_011535963.2 XP_011534265.1 1 546
XM_017011082.1 XP_016866571.1 1 546
GenBank mRNA AB210839.1 - 1 236
AK000293.1 - 1 478
AK225629.1 - 1 507
AK296717.1 - 1 507
AK315804.1 - 1 507
BC029491.1 - 1 516
BC039906.1 - 1 512 6066481
DC341808.1 - 1 527

View Full Product Details