Gene Transcripts
- Gene Symbol
- TGFB1
- Entrez Gene ID
- 7040
- Gene Name
- transforming growth factor beta 1
- Gene Aliases
- CED, DPD1, LAP, TGFB, TGFbeta
- Chromosome Location
- Chr. 19: 41330531 - 41353933 on Build GRCh38
- UniGene ID
- Hs.645227
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
X02812.1 |
- |
7 |
2430 |
|
View Full Product Details