Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010046.2 | NP_034176.1 | 8, 9 | 821 | |
GenBank mRNA | AB057816.1 | - | 8, 9 | 809 | |
AF078752.1 | - | 8, 9 | 879 | ||
AK215091.1 | - | 2, 3 | 103 | ||
BC003717.1 | - | 8, 9 | 821 | 3486724 |